Thermo Scientific Chemicals

Indium(III) phosphate, 98%, Thermo Scientific Chemicals

Catalog number: A18281.06
5 g, Each
Thermo Scientific Chemicals

Indium(III) phosphate, 98%, Thermo Scientific Chemicals

Catalog number: A18281.06
5 g, Each
Quantity
Catalog number: A18281.06
also known as A18281-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14693-82-4
IUPAC Name
indium(3+) phosphate
Molecular Formula
InO4P
InChI Key
UJXZVRRCKFUQKG-UHFFFAOYSA-K
SMILES
[In+3].[O-]P([O-])([O-])=O
Form
Powder
Elemental Analysis
In: 53.36-56.10% (Theory: 54.73%)
Appearance (Color)
White
Assay (special)
≥97.5 to ≤102.5% (Based on elemental In)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text