Thermo Scientific Chemicals

Stearic acid hydrazide, 97%, Thermo Scientific Chemicals

Catalog number: A18291.18
50 g, Each
Thermo Scientific Chemicals

Stearic acid hydrazide, 97%, Thermo Scientific Chemicals

Catalog number: A18291.18
50 g, Each
Quantity
Catalog number: A18291.18
also known as A18291-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4130-54-5
IUPAC Name
octadecanehydrazide
Molecular Formula
C18H38N2O
InChI Key
BYTFESSQUGDMQQ-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCC(=O)NN
Form
Crystalline powder or powder
Melting Point (clear melt)
104-111?C
Appearance (Color)
White to cream
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text