Thermo Scientific Chemicals

Rhodanine, 98+%, Thermo Scientific Chemicals

Catalog number: A18293.36
500 g, Each
Thermo Scientific Chemicals

Rhodanine, 98+%, Thermo Scientific Chemicals

Catalog number: A18293.36
500 g, Each
Quantity
Catalog number: A18293.36
also known as A18293-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-84-4
IUPAC Name
2-sulfanylidene-1,3-thiazolidin-4-one
Molecular Formula
C3H3NOS2
InChI Key
KIWUVOGUEXMXSV-UHFFFAOYSA-N
SMILES
O=C1CSC(=S)N1
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to cream to yellow to pale brown
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Assay (HPLC)
≥98.0%

Description

Rhodanine inhibits the multiplication of echovirus 12 and the development of virus-induced morphologic changes. It has been used in tannase assay in cultures of tannin degrading fungi. It possess anticonvulsant, antibacterial, antiviral and antidiabetic activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rhodanine inhibits the multiplication of echovirus 12 and the development of virus-induced morphologic changes. It has been used in tannase assay in cultures of tannin degrading fungi. It possess anticonvulsant, antibacterial, antiviral and antidiabetic activities.

Solubility
Soluble in water (2.25 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text