Thermo Scientific Chemicals

2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide, 99%, Thermo Scientific Chemicals

Catalog number: A18294.06
5 g, Each
Thermo Scientific Chemicals

2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide, 99%, Thermo Scientific Chemicals

Catalog number: A18294.06
5 g, Each
Quantity
Catalog number: A18294.06
also known as A18294-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19805-75-5
IUPAC Name
3-carbamoyl-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrol-1-ium
Molecular Formula
C9H17N2O
InChI Key
ACFYUJLIWIDSFM-UHFFFAOYSA-O
SMILES
CC1(C)[NH2+]C(C)(C)C(=C1)C(N)=O
Assay (HPLC)
≥98.5%
Melting Point
177.5-183.5?C
Appearance (Color)
White to orange-brown
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms

Description

2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide may be used in the synthesis of 2,2,5,5-tetramethyl-3-pyrrolidinecarboxamide and 3-carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy free radical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide may be used in the synthesis of 2,2,5,5-tetramethyl-3-pyrrolidinecarboxamide and 3-carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy free radical.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text