N,N-Dimethyl-4-nitrosoaniline, 98%, Thermo Scientific Chemicals
N,N-Dimethyl-4-nitrosoaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Dimethyl-4-nitrosoaniline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
Catalog number A18301.30
also known as A18301-30
Price (USD)/ Each
224.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
224.00
Add to cart
N,N-Dimethyl-4-nitrosoaniline, 98%, Thermo Scientific Chemicals
Catalog numberA18301.30
Price (USD)/ Each
224.00
-
Add to cart
Chemical Identifiers
CAS138-89-6
IUPAC NameN,N-dimethyl-4-nitrosoaniline
Molecular FormulaC8H10N2O
InChI KeyCMEWLCATCRTSGF-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)N=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green
FormCrystals or powder
Assay (GC)>97.5%
Solution Test1% w/v in ethanol : Clear
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only