Thermo Scientific Chemicals

(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate, 98%, Thermo Scientific Chemicals

Catalog number: A18305.06
5 g, Each
Thermo Scientific Chemicals

(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate, 98%, Thermo Scientific Chemicals

Catalog number: A18305.06
5 g, Each
Quantity
Catalog number: A18305.06
also known as A18305-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
52186-89-7
IUPAC Name
[1-(ethoxycarbonyl)cyclopropyl]triphenylphosphanium; tetrafluoroboranuide
Molecular Formula
C24H24BF4O2P
InChI Key
RGJYRMUGSAFITK-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.CCOC(=O)C1(CC1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to pale cream
Form
crystalline powder
Assay (unspecified)
> 97.5% by Carbon and Phosphorus EA
Elemental Analysis
Carbon C: 60.81 - 63.92% (theory 62.36%)
Elemental Analysis
Phosphorus P: 6.54 - 6.87% (theory 6.70%)

Description

(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text