Chemical Identifiers
CAS85-86-9
IUPAC Name(1Z)-1-{2-[4-(2-phenyldiazen-1-yl)phenyl]hydrazin-1-ylidene}-1,2-dihydronaphthalen-2-one
Molecular FormulaC22H16N4O
InChI KeyHTPQPMPFXUWUOT-SRURNFRUSA-N
SMILESO=C1C=CC2=CC=CC=C2\C1=N\NC1=CC=C(C=C1)N=NC1=CC=CC=C1
View more