Nitrilotriacetic acid trisodium salt monohydrate, 98+%, Thermo Scientific Chemicals
Nitrilotriacetic acid trisodium salt monohydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nitrilotriacetic acid trisodium salt monohydrate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
Catalog number A18348.0E
also known as A18348-0E
Price (USD)/ Each
198.00
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
198.00
Add to cart
Nitrilotriacetic acid trisodium salt monohydrate, 98+%, Thermo Scientific Chemicals
Catalog numberA18348.0E
Price (USD)/ Each
198.00
-
Add to cart
Chemical Identifiers
CAS18662-53-8
IUPAC Nametrisodium 2-[bis(carboxylatomethyl)amino]acetate hydrate
Molecular FormulaC6H8NNa3O7
InChI KeyHERBOKBJKVUALN-UHFFFAOYSA-K
SMILESO.[Na+].[Na+].[Na+].[O-]C(=O)CN(CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (ex Sulphated Ash)≥98.0%
FormCrystals or powder or crystalline powder or fused solid
Water Content (Karl Fischer Titration)3.38-9.51% (0.5-1.5 waters)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only