2-Mercaptobenzimidazole, 97%
2-Mercaptobenzimidazole, 97%
2-Mercaptobenzimidazole, 97%
2-Mercaptobenzimidazole, 97%
Thermo Scientific Chemicals

2-Mercaptobenzimidazole, 97%

CAS: 583-39-1 | C7H6N2S | 150.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A18350.36
also known as A18350-36
Price (USD)
66.65
Online Exclusive
74.40
Save 7.75 (10%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
66.65
Online Exclusive
74.40
Save 7.75 (10%)
Each
Add to cart
Specifications
Beilstein119867
CAS583-39-1
Chemical Name or Material2-Mercaptobenzimidazole
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Flash Point>250°C (487°F)
View more
Ask our AI about this Product
2-Mercaptobenzimidazole is used as an antidegradant. It is utilized for protecting rubber from oxidation. It is also employed as an intermediate in the preparation of rabeprazole. Further, it finds application in anti-leprosy drugs in medicine. In addition to this, it is used as a copper plating brightener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercaptobenzimidazole is used as an antidegradant. It is utilized for protecting rubber from oxidation. It is also employed as an intermediate in the preparation of rabeprazole. Further, it finds application in anti-leprosy drugs in medicine. In addition to this, it is used as a copper plating brightener.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Zhong, B.; Jia, Z.; Luo, Y.; Guo, B.; Jia, D. Preparation of halloysite nanotubes supported 2-mercaptobenzimidazole and its application in natural rubber. Composites Part A 2015, 73, 63-71.
  2. Žerjav, G.; Milošev, I. Protection of copper against corrosion in simulated urban rain by the combined action of benzotriazole, 2-mercaptobenzimidazole and stearic acid. Corros. Sci. 2015, 98, 180-191.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
P05F008Certificate of AnalysisFeb 15, 2023A18350.36, A18350.0E, A18350.22, ALF-A18350-0E, ALF-A18350-22, ALF-A18350-36
Z10A006Certificate of AnalysisFeb 15, 2023A18350.36, A18350.0E, A18350.22, ALF-A18350-0E, ALF-A18350-22, ALF-A18350-36
M13C018Certificate of AnalysisNov 23, 2021A18350.0E, A18350.36, A18350.22
10138978Certificate of AnalysisNov 23, 2021A18350.0E, A18350.36, A18350.22
10137724Certificate of AnalysisNov 23, 2021A18350.0E, A18350.36, A18350.22
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link