trans-2,cis-6-Nonadienal, 90+% (major isomer), remainder mainly trans,trans-isomer, Thermo Scientific Chemicals
trans-2,cis-6-Nonadienal, 90+% (major isomer), remainder mainly trans,trans-isomer, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-2,cis-6-Nonadienal, 90+% (major isomer), remainder mainly trans,trans-isomer, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number A18356.14
also known as A18356-14
Price (USD)/ Each
342.65
Online exclusive
381.00 
Save 38.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
342.65
Online exclusive
381.00 
Save 38.35 (10%)
Add to cart
trans-2,cis-6-Nonadienal, 90+% (major isomer), remainder mainly trans,trans-isomer, Thermo Scientific Chemicals
Catalog numberA18356.14
Price (USD)/ Each
342.65
Online exclusive
381.00 
Save 38.35 (10%)
-
Add to cart
Chemical Identifiers
CAS557-48-2
IUPAC Name(2E,6Z)-nona-2,6-dienal
Molecular FormulaC9H14O
InChI KeyHZYHMHHBBBSGHB-ODYTWBPASA-N
SMILESCCC=C/CCC=CC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)<1.0%
FormLiquid
Refractive Index1.4700-1.4780 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥90.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only