Ethyl Red
Thermo Scientific Chemicals

Ethyl Red

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number A18367.30
also known as A18367-30
Price (USD)/ Each
410.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
410.00
Add to cart
Ethyl Red
Catalog numberA18367.30
Price (USD)/ Each
410.00
-
Add to cart
Chemical Identifiers
CAS76058-33-8
IUPAC Name2-{2-[4-(diethylamino)phenyl]diazen-1-yl}benzoate
Molecular FormulaC17H18N3O2
InChI KeyHBRCDTRQDHMTDA-UHFFFAOYSA-M
SMILESCCN(CC)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red to dark purple powder
FormPowder
Color ChangeRed @ pH 4.5 to yellow @ pH 6.5
lambda-max (UV-Vis)450nm ± 5nm (0.1M NaOH)
Solution Test0.1% in ethanol: clear
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only