Trifluoroacetaldehyde methyl hemiacetal, tech. 90%, Thermo Scientific Chemicals
Trifluoroacetaldehyde methyl hemiacetal, tech. 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trifluoroacetaldehyde methyl hemiacetal, tech. 90%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number A18369.22
also known as A18369-22
Price (USD)/ Each
442.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
442.00
Add to cart
Trifluoroacetaldehyde methyl hemiacetal, tech. 90%, Thermo Scientific Chemicals
Catalog numberA18369.22
Price (USD)/ Each
442.00
-
Add to cart
Chemical Identifiers
CAS431-46-9
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3260-1.3300 @ 20?C
Appearance (Color)Clear, colorless
FormLiquid
Proton NMR<10 wt.% methanol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only