Thermo Scientific Chemicals

4,4'-(Hexafluoroisopropylidene)diphenol, 98%, Thermo Scientific Chemicals

Catalog number: A18370.22
100 g, Each
Thermo Scientific Chemicals

4,4'-(Hexafluoroisopropylidene)diphenol, 98%, Thermo Scientific Chemicals

Catalog number: A18370.22
100 g, Each
Quantity
Catalog number: A18370.22
also known as A18370-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1478-61-1
Melting Point (clear melt)
158-164?C
Appearance (Color)
White to cream
Form
Powder
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

4,4'-(Hexafluoroisopropylidene)diphenol is used to cross-link and cure fluoroelastomers. is also used as a monomer in the synthesis of specialty polymers. Polymers containing Bisphenol AF are useful in high temperature composites, in electronic materials and other specialty applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-(Hexafluoroisopropylidene)diphenol is used to cross-link and cure fluoroelastomers. is also used as a monomer in the synthesis of specialty polymers. Polymers containing Bisphenol AF are useful in high temperature composites, in electronic materials and other specialty applications.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text