Thermo Scientific Chemicals

Cobalt(II) acetate tetrahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A18374.14
25 g, Each
Thermo Scientific Chemicals

Cobalt(II) acetate tetrahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A18374.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A18374.14
also known as A18374-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6147-53-1
IUPAC Name
λ²-cobalt(2+) diacetate tetrahydrate
Molecular Formula
C4H14CoO8
InChI Key
ZBYYWKJVSFHYJL-UHFFFAOYSA-L
SMILES
O.O.O.O.[Co++].CC([O-])=O.CC([O-])=O
Appearance (Color)
Dark pink to red-pink
Form
Crystals or crystalline powder
Elemental Analysis
Cobalt: 23.19-24.13%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
28.0-30.5%

Description

Cobalt(II) acetate tetrahydrate is used as an industrial catalyst to harden paints and varnishes, an active catalyst for oxidation and esterification reactions. It is also used to prepare metal complexes having unusual coordination geometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) acetate tetrahydrate is used as an industrial catalyst to harden paints and varnishes, an active catalyst for oxidation and esterification reactions. It is also used to prepare metal complexes having unusual coordination geometry.

Solubility
Soluble in water, alcohol, dilute acids and phenyl acetate.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text