1-Fluoronaphthalene is used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Fluoronaphthalene is used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It is also used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- Wojciech A. Majewski.; David F. Plusquellic.; David W. Pratt. The rotationally resolved fluorescence excitation spectrum of 1-fluoronaphthalene. J. Chem. Phys. 1989, 90 (3), 1362.
- J. Mispelter.; J.-Ph. Grivet.; J.-M. Lhoste. Electron spin resonance of phosphorescent fluorine-substituted aromatics I. 1-Fluoronaphthalene and 2-fluoronaphthalene in durene single crystals. Molecular Physics: An International Journal at the Interface Between Chemistry and Physics. 1971, 21 (6), 999-1013.