Thermo Scientific Chemicals

1-Fluoronaphthalene, 98%, Thermo Scientific Chemicals

Catalog number: A18378.22
100 g, Each
Thermo Scientific Chemicals

1-Fluoronaphthalene, 98%, Thermo Scientific Chemicals

Catalog number: A18378.22
100 g, Each
Quantity
Catalog number: A18378.22
also known as A18378-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
321-38-0
IUPAC Name
1-fluoronaphthalene
Molecular Formula
C10H7F
InChI Key
CWLKTJOTWITYSI-UHFFFAOYSA-N
SMILES
FC1=C2C=CC=CC2=CC=C1
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.5910-1.5950 @ 20?C

Description

1-Fluoronaphthalene is used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoronaphthalene is used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text