Thermo Scientific Chemicals

Sodium hydroxide, flake, 98%, Thermo Scientific Chemicals

Catalog number: A18395.0E
2500 g, Each
Thermo Scientific Chemicals

Sodium hydroxide, flake, 98%, Thermo Scientific Chemicals

Catalog number: A18395.0E
2500 g, Each
Quantity
Catalog number: A18395.0E
also known as A18395-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1310-73-2
IUPAC Name
sodium hydroxide
Molecular Formula
HNaO
InChI Key
HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES
[OH-].[Na+]
Appearance (Color)
White to orange
Form
Flakes
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5% (UK-Sourced Material)
Assay from Supplier's CofA
≥97.5% (US-Sourced Material)
Comment
Material Sourced in UK and US

Description

Sodium hydroxide flake is widely used in a variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as in water treatment plants. It is used in the Bayer process of aluminum manufacture as well as to remove sulfurous impurities from crude oil. It is also used in the manufacture of biodiesel, rayon and cellophane; as a catalyst for the trans esterification of methanol and triglycerides and as paint stripper on wooden objects. It is mainly used for pH control, acid neutralization, off-gas scrubbing and as a catalyst in various industries like pharmaceutical, agrochemical, dyes, paints and polymer. It is also used in refining vegetable oils, rubber reclamation, metal processing, metal degreasing, adhesive preparations, paint removers, disinfectants, rubber latex stabilizers and in the stabilization of sodium hypochlorite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydroxide flake is widely used in a variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as in water treatment plants. It is used in the Bayer process of aluminum manufacture as well as to remove sulfurous impurities from crude oil. It is also used in the manufacture of biodiesel, rayon and cellophane; as a catalyst for the trans esterification of methanol and triglycerides and as paint stripper on wooden objects. It is mainly used for pH control, acid neutralization, off-gas scrubbing and as a catalyst in various industries like pharmaceutical, agrochemical, dyes, paints and polymer. It is also used in refining vegetable oils, rubber reclamation, metal processing, metal degreasing, adhesive preparations, paint removers, disinfectants, rubber latex stabilizers and in the stabilization of sodium hypochlorite.

Notes
Air sensitive and hygroscopic. It is deliquescent and readily absorbs moisture and carbon dioxide in air. Incompatible with strong oxidizing agents, strong acids and organic materials.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text