Thermo Scientific Chemicals

Ethyl 6-bromohexanoate, 97+%, Thermo Scientific Chemicals

Catalog number: A18398.18
50 g, Each
Thermo Scientific Chemicals

Ethyl 6-bromohexanoate, 97+%, Thermo Scientific Chemicals

Catalog number: A18398.18
50 g, Each
Quantity
Catalog number: A18398.18
also known as A18398-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25542-62-5
IUPAC Name
ethyl 6-bromohexanoate
Molecular Formula
C8H15BrO2
InChI Key
DXBULVYHTICWKT-UHFFFAOYSA-N
SMILES
CCOC(=O)CCCCCBr
Refractive Index
1.4575-1.4625 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.0%

Description

Ethyl 6-bromohexanoate is used in the preparation of various carnitine derivatives in a carnitine transporter study. It is also used for alkylation of pentane-2,4-dione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 6-bromohexanoate is used in the preparation of various carnitine derivatives in a carnitine transporter study. It is also used for alkylation of pentane-2,4-dione.

Solubility
Soluble in water (water, 64.76 mg/L @ 25°C (est.)).

Notes
Store away from strong oxidizing agents, strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text