Thermo Scientific Chemicals

7-Amino-4-(trifluoromethyl)coumarin, 97%, Thermo Scientific Chemicals

Catalog number: A18400.01
0.25 g, Each
Thermo Scientific Chemicals

7-Amino-4-(trifluoromethyl)coumarin, 97%, Thermo Scientific Chemicals

Catalog number: A18400.01
0.25 g, Each
Quantity
Catalog number: A18400.01
also known as A18400-01
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
53518-15-3
IUPAC Name
7-amino-4-(trifluoromethyl)-2H-chromen-2-one
Molecular Formula
C10H6F3NO2
InChI Key
JBNOVHJXQSHGRL-UHFFFAOYSA-N
SMILES
NC1=CC=C2C(OC(=O)C=C2C(F)(F)F)=C1
Melting Point (clear melt)
218.0-227.0?C
Appearance (Color)
Yellow
Assay (HPLC)
≥98.5%
Form
Crystals or powder or crystalline powder

Description

7-Amino-4-(trifluoromethyl)coumarin is used in the synthesis of substrates for the fluorometric assay of proteolytic enzymes in biological fluids. It is suitable as laser dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Amino-4-(trifluoromethyl)coumarin is used in the synthesis of substrates for the fluorometric assay of proteolytic enzymes in biological fluids. It is suitable as laser dye.

Solubility
Soluble in dimethyl sulfoxide (25 mg/ml), dimethyl formamide (25 mg/ml), and methanol (25 mg/ml). Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text