Thermo Scientific Chemicals

Dibenzosuberone, 97%, Thermo Scientific Chemicals

Catalog number: A18405.22
100 g, Each
Thermo Scientific Chemicals

Dibenzosuberone, 97%, Thermo Scientific Chemicals

Catalog number: A18405.22
100 g, Each
Quantity
Catalog number: A18405.22
also known as A18405-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1210-35-1
Assay (GC)
≥96.0%
Appearance (Color)
Clear pale yellow to yellow
Form
Liquid
Identification (FTIR)
Conforms

Description

Dibenzosuberone is a potential antidepressant compound. It is used in the synthesis of amitriptyline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzosuberone is a potential antidepressant compound. It is used in the synthesis of amitriptyline.

Solubility
Soluble in water (0.03g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text