Thermo Scientific Chemicals

1,2-Propanediol, 98+%, Thermo Scientific Chemicals

Catalog number: A18406.36
500 g, Each
Thermo Scientific Chemicals

1,2-Propanediol, 98+%, Thermo Scientific Chemicals

Catalog number: A18406.36
500 g, Each
Quantity
Catalog number: A18406.36
also known as A18406-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57-55-6
IUPAC Name
propane-1,2-diol
Molecular Formula
C3H8O2
InChI Key
DNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILES
CC(O)CO
Refractive Index
1.4305-1.4345 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms

Description

1,2-Propanediol is used in antifreeze and deicing solutions for cars, boats and aircraft. It is also used to create harmless, artificial smoke for training and theatrical purposes. It is used as a humectant solvent and preservative in food. It acts as a solvent for paints and plastics. Further, it is used in the manufacturing of lactic acid by green catalytic oxidation, propanal and propylene carbonate. It is also employed in veterinary medicine as an oral treatment for hyperketonaemia in ruminants. In addition to this, it reacts with propylene oxide to prepare oligomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Propanediol is used in antifreeze and deicing solutions for cars, boats and aircraft. It is also used to create harmless, artificial smoke for training and theatrical purposes. It is used as a humectant solvent and preservative in food. It acts as a solvent for paints and plastics. Further, it is used in the manufacturing of lactic acid by green catalytic oxidation, propanal and propylene carbonate. It is also employed in veterinary medicine as an oral treatment for hyperketonaemia in ruminants. In addition to this, it reacts with propylene oxide to prepare oligomers.

Solubility
Miscible with water, benzene, acetone, chloroform and ether. Immiscible with fixed oils.

Notes
Hygroscopic. Incompatible with acid chlorides, acid anhydrides, oxidizing agents, chloroformates and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text