1,4-Bis(trifluoromethyl)benzene, 99%, Thermo Scientific Chemicals
1,4-Bis(trifluoromethyl)benzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Bis(trifluoromethyl)benzene, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 g
Catalog number A18411.22
also known as A18411-22
Price (USD)/ Each
267.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
267.00
Add to cart
1,4-Bis(trifluoromethyl)benzene, 99%, Thermo Scientific Chemicals
Catalog numberA18411.22
Price (USD)/ Each
267.00
-
Add to cart
Chemical Identifiers
CAS433-19-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Refractive Index1.3780-1.3810 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only