Thermo Scientific Chemicals

(Iodomethyl)trimethylammonium iodide, 98+%, Thermo Scientific Chemicals

Catalog number: A18424.14
25 g, Each
Thermo Scientific Chemicals

(Iodomethyl)trimethylammonium iodide, 98+%, Thermo Scientific Chemicals

Catalog number: A18424.14
25 g, Each
Quantity
Catalog number: A18424.14
also known as A18424-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39741-91-8
IUPAC Name
(iodomethyl)trimethylazanium iodide
Molecular Formula
C4H11I2N
InChI Key
IFVIWWQMNYZYFQ-UHFFFAOYSA-M
SMILES
[I-].C[N+](C)(C)CI
Assay (Titration ex Iodide)
≥98.0 to ≤102.0%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤1.0%
Appearance (Color)
White to pale cream or pale yellow
Form
Crystalline powder or powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text