Thermo Scientific Chemicals

(3-Glycidoxypropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: A18431.22
100 g, Each
Thermo Scientific Chemicals

(3-Glycidoxypropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: A18431.22
100 g, Each
Quantity
Catalog number: A18431.22
also known as A18431-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2530-83-8
IUPAC Name
trimethoxy({3-[(oxiran-2-yl)methoxy]propyl})silane
Molecular Formula
C9H20O5Si
InChI Key
BPSIOYPQMFLKFR-UHFFFAOYNA-N
SMILES
CO[Si](CCCOCC1CO1)(OC)OC
Refractive Index
1.4265-1.4315 @ 20°C
Solution Test
Solubility: Passes Test (U.S. specification)
Viscosity
2.9-3.2 cSt (U.S. specification)
Appearance (Color)
Clear colorless
Form
Liquid

Description

(3-Glycidoxypropyl)trimethoxysilane is used in the production of carbon steel and its pre treatment favors the dry and wet adhesion of epoxy coating. It is also used to prepare epoxy-functionalized silica nanoparticles, which gives a reactive surface for one-step and high-density protein immobilization. It serves as a coupling agent and adhesion promoter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Glycidoxypropyl)trimethoxysilane is used in the production of carbon steel and its pre treatment favors the dry and wet adhesion of epoxy coating. It is also used to prepare epoxy-functionalized silica nanoparticles, which gives a reactive surface for one-step and high-density protein immobilization. It serves as a coupling agent and adhesion promoter.

Solubility
Miscible with alcohols, ketones and aliphatic or aromatic hydrocarbons. Immiscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text