Germanium(IV) methoxide, 98%, Thermo Scientific Chemicals
Germanium(IV) methoxide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Germanium(IV) methoxide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A18437.14
also known as A18437-14
Price (USD)/ Each
681.65
Online exclusive
757.00 
Save 75.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
681.65
Online exclusive
757.00 
Save 75.35 (10%)
Add to cart
Germanium(IV) methoxide, 98%, Thermo Scientific Chemicals
Catalog numberA18437.14
Price (USD)/ Each
681.65
Online exclusive
757.00 
Save 75.35 (10%)
-
Add to cart
Chemical Identifiers
CAS992-91-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
FormLiquid
Assay (unspecified)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only