Thermo Scientific Chemicals

Cyclohexylamine hydrobromide, 98%, Thermo Scientific Chemicals

Catalog number: A18439.36
500 g, Each
Thermo Scientific Chemicals

Cyclohexylamine hydrobromide, 98%, Thermo Scientific Chemicals

Catalog number: A18439.36
500 g, Each
Quantity
Catalog number: A18439.36
also known as A18439-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26227-54-3
IUPAC Name
hydrogen cyclohexanamine bromide
Molecular Formula
C6H14BrN
InChI Key
QOHWJRRXQPGIQW-UHFFFAOYSA-N
SMILES
[H+].[Br-].NC1CCCCC1
Appearance (Color)
White to pale cream
Assay (Titration ex Bromide)
≥97.5 to ≤102.5%
Form
Crystalline powder
Water Content (Karl Fischer Titration)
<1.0%
Melting Point (clear melt)
196-203?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text