Thermo Scientific Chemicals

Nickel ammonium sulfate hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A18441.22
100 g, Each
Thermo Scientific Chemicals

Nickel ammonium sulfate hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A18441.22
100 g, Each
Quantity
Catalog number: A18441.22
also known as A18441-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7785-20-8
Appearance (Color)
Blue to green
Form
Powder or Crystals
Assay (unspecified)
>97.5 to <102.5% (as hexahydrate)

Description

Nickel ammonium sulfate hexahydrate is used as a nickel electrolyte for electroplating and as an analytical reagent. It is also used as a soil fertilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nickel ammonium sulfate hexahydrate is used as a nickel electrolyte for electroplating and as an analytical reagent. It is also used as a soil fertilizer.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text