Nickel ammonium sulfate hexahydrate is used as a nickel electrolyte for electroplating and as an analytical reagent. It is also used as a soil fertilizer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nickel ammonium sulfate hexahydrate is used as a nickel electrolyte for electroplating and as an analytical reagent. It is also used as a soil fertilizer.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
Meena, K.; Muthu, K.; Rajasekar, M.; Bhagavannarayana, G.; Meenakshisundaram, S. P.; Mojumdar, S. C. Urea/thiourea induced crystal growth of ammonium nickel sulfate hexahydrate and characterization studies. J. Therm. Anal. Calorim. 2015, 119 (2), 963-968.
Singh, S.; Patel, A.; Prakashan, P. One pot oxidative esterification of aldehyde over recyclable cesium salt of nickel substituted phosphotungstate. Appl. Catal., A 2015, 505, 131-140.