Thermo Scientific Chemicals

Diisopropanolamine, 98+%, sum of isomers, Thermo Scientific Chemicals

Catalog number: A18451.0B
1000 g, Each
Thermo Scientific Chemicals

Diisopropanolamine, 98+%, sum of isomers, Thermo Scientific Chemicals

Catalog number: A18451.0B
1000 g, Each
Quantity
Catalog number: A18451.0B
also known as A18451-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-97-4
Appearance (Color)
White
Form
Crystalline or fused solid
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
<1.0%

Description

Bis(2-hydroxypropyl)amine (Diisopropanolamine) is used to study its effects upon choline uptake and phospholipid synthesis in Chinese hamster ovary (CHO) cells. It is a versatile chemical that is widely used as an emulsifier, stabilizer, surfactant, and chemical building block. It can neutralize pH, act as a buffer (stabilize pH), or add basicity (alkalinity) to a solution. Major applications include personal care products, metalworking products, and natural gas purification. Diisopropanolamine is also used in coatings, paint strippers, paper, photographic intermediates, plastics, polyurethanes, and textiles. It is frequently used to neutralize fatty acids and sulfonic acidbased surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-hydroxypropyl)amine (Diisopropanolamine) is used to study its effects upon choline uptake and phospholipid synthesis in Chinese hamster ovary (CHO) cells. It is a versatile chemical that is widely used as an emulsifier, stabilizer, surfactant, and chemical building block. It can neutralize pH, act as a buffer (stabilize pH), or add basicity (alkalinity) to a solution. Major applications include personal care products, metalworking products, and natural gas purification. Diisopropanolamine is also used in coatings, paint strippers, paper, photographic intermediates, plastics, polyurethanes, and textiles. It is frequently used to neutralize fatty acids and sulfonic acidbased surfactants.

Solubility
Soluble in water 20°C 870g/L. Miscible in alcohol.

Notes
Air Sensitive & Hygroscopic. Store away from oxidizing agents, air, water/moisture and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text