Thermo Scientific Chemicals

1,5-Dibromopentane, 98%, Thermo Scientific Chemicals

Catalog number: A18465.36
500 g, Each
Thermo Scientific Chemicals

1,5-Dibromopentane, 98%, Thermo Scientific Chemicals

Catalog number: A18465.36
500 g, Each
Quantity
Catalog number: A18465.36
also known as A18465-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-24-0
IUPAC Name
1,5-dibromopentane
Molecular Formula
C5H10Br2
InChI Key
IBODDUNKEPPBKW-UHFFFAOYSA-N
SMILES
BrCCCCCBr
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Refractive Index
1.5090-1.5145 @ 20?C
Form
Liquid

Description

1,5-Dibromopentane is used in the preparation of 1,5-di-Grignard reagent and novel spirocyclic pyrrolidones. It acts as a growth supplement for Yarrowia lipolytica 3589, a tropical marine yeast. It is involved in the synthesis of N-alkylated piperidine as well as tetrydrothiopyran (thiane) by reaction with primary amine and sodium sulfide respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dibromopentane is used in the preparation of 1,5-di-Grignard reagent and novel spirocyclic pyrrolidones. It acts as a growth supplement for Yarrowia lipolytica 3589, a tropical marine yeast. It is involved in the synthesis of N-alkylated piperidine as well as tetrydrothiopyran (thiane) by reaction with primary amine and sodium sulfide respectively.

Solubility
Miscible with water, slightly miscible with chloroform and benzene.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text