Thermo Scientific Chemicals

Phthalimidoacetaldehyde diethyl acetal, 99%, Thermo Scientific Chemicals

Catalog number: A18491.14
25 g, Each
Thermo Scientific Chemicals

Phthalimidoacetaldehyde diethyl acetal, 99%, Thermo Scientific Chemicals

Catalog number: A18491.14
25 g, Each
Quantity
Catalog number: A18491.14
also known as A18491-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78902-09-7
IUPAC Name
2-(2,2-diethoxyethyl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular Formula
C14H17NO4
InChI Key
GEFXJJJQUSEHLV-UHFFFAOYSA-N
SMILES
CCOC(CN1C(=O)C2=CC=CC=C2C1=O)OCC
Appearance (Color)
White to pale yellow
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%
Melting Point (clear melt)
70.0-76.0?C

Description

Phthalimidoacetaldehyde diethyl acetal is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalimidoacetaldehyde diethyl acetal is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text