D-Panthenol, 98+%, Thermo Scientific Chemicals
D-Panthenol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Panthenol, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A18499.36
also known as A18499-36
Price (USD)/ Each
187.65
Online exclusive
208.00 
Save 20.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
187.65
Online exclusive
208.00 
Save 20.35 (10%)
Add to cart
D-Panthenol, 98+%, Thermo Scientific Chemicals
Catalog numberA18499.36
Price (USD)/ Each
187.65
Online exclusive
208.00 
Save 20.35 (10%)
-
Add to cart
Chemical Identifiers
CAS81-13-0
IUPAC Name(2R)-2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
Molecular FormulaC9H19NO4
InChI KeySNPLKNRPJHDVJA-ZETCQYMHSA-N
SMILESCC(C)(CO)[C@@H](O)C(=O)NCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Water Content (Karl Fischer Titration)≤1.0%
Refractive Index1.4950-1.5020 @ 20°C
FormViscous liquid
Identification (FTIR)Conforms
View more
D-Panthenol is used as precursors in the biosynthesis of coenzyme A. D-Panthenol and D-Pantothenic acid are used in a variety of skin protection products. D-pantothenic acid have a role in controlling keratinocyte proliferation and differentiation. D-Panthenol is used for studies of skin protection emulsions from UV irradiation and as a humectant. It is also used in creams, lotions, shampoos, conditioners and body washes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Panthenol is used as precursors in the biosynthesis of coenzyme A. D-Panthenol and D-Pantothenic acid are used in a variety of skin protection products. D-pantothenic acid have a role in controlling keratinocyte proliferation and differentiation. D-Panthenol is used for studies of skin protection emulsions from UV irradiation and as a humectant. It is also used in creams, lotions, shampoos, conditioners and body washes.

Solubility
Fully miscible in water and soluble in alcohol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
RUO – Research Use Only

General References:

  1. Lai-Hao Wang.; Shih-Wen Tseng. Direct determination of d-panthenol and salt of pantothenic acid in cosmetic and pharmaceutical preparations by differential pulse voltammetry. Analytica Chimica Acta. 2001, 432 (1), 39-48.
  2. Rajeshwari V. Hosahalli.; Anita P. Savanur.; Sharanappa T. Nandibewoor.; Shivamurti A. Chimatadar. Kinetics and mechanism of uncatalysed and ruthenium(III)-catalysed oxidation of d-panthenol by alkaline permanganate. Transition Metal Chemistry. 2010, 35 (2), 237-246.