Thermo Scientific Chemicals

2,4-Di-tert-butylphenol, 97%, Thermo Scientific Chemicals

Catalog number: A18509.30
250 g, Each
Thermo Scientific Chemicals

2,4-Di-tert-butylphenol, 97%, Thermo Scientific Chemicals

Catalog number: A18509.30
250 g, Each
Quantity
Catalog number: A18509.30
also known as A18509-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-76-4
IUPAC Name
2,4-di-tert-butylphenol
Molecular Formula
C14H22O
InChI Key
ICKWICRCANNIBI-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC=C(O)C(=C1)C(C)(C)C
Assay (GC)
≥96.0% (non-U.S. specification)
Assay from Suppliers CofA
≥96.0% (U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated
Melting Point (clear melt)
52.0-58.0°C (non-U.S. specification)
Appearance (Color)
Yellow to brown

Description

2,4-Di-tert-butylphenol is used to study its alkylation reaction with cinnamyl alcohol in the presence of Al-substituted disordered hexagonal molecular silica catalysts. It is a raw material for phosphite anti-oxidants. The antioxidants are primarily used in the manufacture of polyolefins. It is also used as an intermediate for the preparation of UV stabilizers and in manufacturing of pharmaceuticals and fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Di-tert-butylphenol is used to study its alkylation reaction with cinnamyl alcohol in the presence of Al-substituted disordered hexagonal molecular silica catalysts. It is a raw material for phosphite anti-oxidants. The antioxidants are primarily used in the manufacture of polyolefins. It is also used as an intermediate for the preparation of UV stabilizers and in manufacturing of pharmaceuticals and fragrances.

Solubility
Soluble in methanol, ether, acetone, chloroform. 2,4-Di-tert-butylphenol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text