Thermo Scientific Chemicals

4-Nitrobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A18512.14
25 g, Each
Thermo Scientific Chemicals

4-Nitrobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A18512.14
25 g, Each
Quantity
Catalog number: A18512.14
also known as A18512-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-74-8
Form
Powder or crystalline powder
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Assay (GC)
≥96.0%
Melting Point (clear melt)
75-80?C
Solution Test
10% in DMF: Clear

Description

4-Nitrobenzenesulfonyl chloride is used as an intermediate for the synthesis of pharmaceuticals like foscmprenavir, darunavir and amprenavir. It is involved in the preparation of N-nosyl-alpha-amino acid for peptide synthesis. It is also used for stereoselective α-glucosylation of partially protected carbohydrates in association with silver tri-fluoromethanesulfonate, N,N-dimethylacetamide and triethylamine. It is widely used for the synthesis of hydroxylamines of sulfadiazine, sulfamethoxazole and [Cu(bipy)2Cl](nbs) (1) (bipy = 2,2'-bipyridine, nbs = 4-nitrobenzenesulfonate).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzenesulfonyl chloride is used as an intermediate for the synthesis of pharmaceuticals like foscmprenavir, darunavir and amprenavir. It is involved in the preparation of N-nosyl-alpha-amino acid for peptide synthesis. It is also used for stereoselective α-glucosylation of partially protected carbohydrates in association with silver tri-fluoromethanesulfonate, N,N-dimethylacetamide and triethylamine. It is widely used for the synthesis of hydroxylamines of sulfadiazine, sulfamethoxazole and [Cu(bipy)2Cl](nbs) (1) (bipy = 2,2′-bipyridine, nbs = 4-nitrobenzenesulfonate).

Solubility
Soluble in toluene, tetrahydrofuran, methylene chloride and ethyl acetate. Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text