Thermo Scientific Chemicals

Manganese(II) nitrate tetrahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A18521.30
250 g, Each
Thermo Scientific Chemicals

Manganese(II) nitrate tetrahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A18521.30
250 g, Each
Quantity
Catalog number: A18521.30
also known as A18521-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20694-39-7
IUPAC Name
manganese(2+) tetrahydrate dinitrate
Molecular Formula
H8MnN2O10
InChI Key
ALIMWUQMDCBYFM-UHFFFAOYSA-N
SMILES
O.O.O.O.[Mn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
Pink
Trace Metal
Zinc (Zn) : ≤ 0.01%
Form
Crystalline powder or fused solid or lumps or chunks
Complexometric Titration
≥97.5 to ≤102.5%
Chloride content
≤ 0.005%

Description

Manganese(II) nitrate tetrahydrate used as porcelain colorants. It is also used as a precursor for manganese dioxide and other manganese compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) nitrate tetrahydrate used as porcelain colorants. It is also used as a precursor for manganese dioxide and other manganese compounds.

Solubility
Soluble in ethanol and water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong reducing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text