1-Acetyl-2-naphthol, 99%, Thermo Scientific Chemicals
1-Acetyl-2-naphthol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Acetyl-2-naphthol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A18537.06
also known as A18537-06
Price (USD)/ Each
26.20
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
26.20
Add to cart
1-Acetyl-2-naphthol, 99%, Thermo Scientific Chemicals
Catalog numberA18537.06
Price (USD)/ Each
26.20
-
Add to cart
Chemical Identifiers
CAS574-19-6
IUPAC Name1-(2-hydroxynaphthalen-1-yl)ethan-1-one
Molecular FormulaC12H10O2
InChI KeyVUIOUIWZVKVFCI-UHFFFAOYSA-N
SMILESCC(=O)C1=C2C=CC=CC2=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)White to cream or pale yellow
FormCrystals or powder or crystalline powder
Melting Point (clear melt)61.0-67.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only