Thermo Scientific Chemicals

Methyl 2-chloropropionate, 97%, Thermo Scientific Chemicals

Catalog number: A18541.30
250 g, Each
Thermo Scientific Chemicals

Methyl 2-chloropropionate, 97%, Thermo Scientific Chemicals

Catalog number: A18541.30
250 g, Each
Quantity
Catalog number: A18541.30
also known as A18541-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17639-93-9
IUPAC Name
methyl 2-chloropropanoate
Molecular Formula
C4H7ClO2
InChI Key
JLEJCNOTNLZCHQ-UHFFFAOYNA-N
SMILES
COC(=O)C(C)Cl
Appearance (Color)
Clear, colourless
Form
liquid
Assay (GC)
>96.0%
Refractive Index
1.4140 - 1.4200

Description

Methyl 2-chloropropionate is used as initiator during the synthesis of acrylamide homopolymers and acrylamide-N-isopropylacrylamide block copolymers. It is also used to produce 2-thiophen-2-yl-propionic acid methyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-chloropropionate is used as initiator during the synthesis of acrylamide homopolymers and acrylamide-N-isopropylacrylamide block copolymers. It is also used to produce 2-thiophen-2-yl-propionic acid methyl ester.

Solubility
Insoluble in water

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text