Thermo Scientific Chemicals

alpha-Naphthoflavone, 97%, Thermo Scientific Chemicals

Catalog number: A18542.06
5 g, Each
Thermo Scientific Chemicals

alpha-Naphthoflavone, 97%, Thermo Scientific Chemicals

Catalog number: A18542.06
5 g, Each
Quantity
Catalog number: A18542.06
also known as A18542-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
604-59-1
IUPAC Name
2-phenyl-4H-benzo[h]chromen-4-one
Molecular Formula
C19H12O2
InChI Key
VFMMPHCGEFXGIP-UHFFFAOYSA-N
SMILES
O=C1C=C(OC2=C1C=CC1=CC=CC=C21)C1=CC=CC=C1
Appearance (Color)
White to pale cream
Form
Powder
Assay (HPLC)
≥96.0%
Melting Point (clear melt)
151.0-161.0?C

Description

An aryl hydrocarbon receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text