Thermo Scientific Chemicals

Salbutamol hemisulfate, 98+%, Thermo Scientific Chemicals

Catalog number: A18544.06
5 g, Each
Thermo Scientific Chemicals

Salbutamol hemisulfate, 98+%, Thermo Scientific Chemicals

Catalog number: A18544.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A18544.06
also known as A18544-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51022-70-9
IUPAC Name
bis(4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol); sulfuric acid
Molecular Formula
C26H44N2O10S
InChI Key
BNPSSFBOAGDEEL-UHFFFAOYNA-N
SMILES
OS(O)(=O)=O.CC(C)(C)NCC(O)C1=CC=C(O)C(CO)=C1.CC(C)(C)NCC(O)C1=CC=C(O)C(CO)=C1
Appearance (Color)
White to pale cream
Form
Powder
Assay (HPLC)
≥98.0%

Description

Salbutamol sulfate is used as standard in analysis of fortified meat for LABA content using ELISA kits. It is also used to study the response of prostaglandin 2α to various β-adrenergic agonists in cultured bovine endometrial cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Salbutamol sulfate is used as standard in analysis of fortified meat for LABA content using ELISA kits. It is also used to study the response of prostaglandin 2α to various β-adrenergic agonists in cultured bovine endometrial cells.

Solubility
Soluble in water and 1M sodium hydroxide (50 mg/ml).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text