1,10-Phenanthroline-2,9-dicarboxylic acid hydrate, 97%, Thermo Scientific Chemicals
1,10-Phenanthroline-2,9-dicarboxylic acid hydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,10-Phenanthroline-2,9-dicarboxylic acid hydrate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
0.25 g
Catalog number A18548.03
also known as A18548-03
Price (USD)/ Each
471.65
Online exclusive
524.00 
Save 52.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
471.65
Online exclusive
524.00 
Save 52.35 (10%)
Add to cart
1,10-Phenanthroline-2,9-dicarboxylic acid hydrate, 97%, Thermo Scientific Chemicals
Catalog numberA18548.03
Price (USD)/ Each
471.65
Online exclusive
524.00 
Save 52.35 (10%)
-
Add to cart
Chemical Identifiers
CAS57709-61-2
IUPAC Name1,10-phenanthroline-2,9-dicarboxylic acid
Molecular FormulaC14H8N2O4
InChI KeyFXSVCROWUPWXBP-UHFFFAOYSA-N
SMILESOC(=O)C1=NC2=C(C=CC3=C2N=C(C=C3)C(O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale green
FormPowder
Assay (HPLC)≥96.0%
Loss on Drying≤14.38% (105?C/constant weight)
1,10-Phenanthroline-2,9-dicarboxylic acid hydrate is used to produce 1,10-phenanthroline-2,9-dicarbonyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Phenanthroline-2,9-dicarboxylic acid hydrate is used to produce 1,10-phenanthroline-2,9-dicarbonyl chloride.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, bases and reducing agents.
RUO – Research Use Only