Thermo Scientific Chemicals

1,2-Cyclohexanediol, cis + trans, 98%, Thermo Scientific Chemicals

Catalog number: A18572.18
50 g, Each
Thermo Scientific Chemicals

1,2-Cyclohexanediol, cis + trans, 98%, Thermo Scientific Chemicals

Catalog number: A18572.18
50 g, Each
Quantity
Catalog number: A18572.18
also known as A18572-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
931-17-9
IUPAC Name
cyclohexane-1,2-diol
Molecular Formula
C6H12O2
InChI Key
PFURGBBHAOXLIO-UHFFFAOYNA-N
SMILES
OC1CCCCC1O
Identification (FTIR)
Conforms
Melting Point (clear melt)
72.0-79.0?C
Appearance (Color)
White to pale cream to pale brown
Form
Crystals or powder or crystalline powder or lumps
Assay (GC)
≥97.5%

Description

1,2-Cyclohexanediol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Cyclohexanediol is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text