Thermo Scientific Chemicals

Field's Stain B

Catalog number: A18578.09
10g, Each
Thermo Scientific Chemicals

Field's Stain B

Catalog number: A18578.09
10g, Each
Quantity
Catalog number: A18578.09
also known as A18578-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17372-87-1
IUPAC Name
disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular Formula
C20H6Br4Na2O5
InChI Key
SEACYXSIPDVVMV-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Br)C(=O)C(Br)=C2OC2=C(Br)C([O-])=C(Br)C=C12
Appearance (Color)
Pink to red/dark red
lambda-max (UV-Vis)
(0.1% w/v in water) : 510-525nm
Specific Absorbance (E 1%/1cm)
350-400 (@ lambda max)
Form
Crystals, powder or crystalline powder
Solution Test
(0.1% w/v in water) : Clear solution

Description

Used to stain blood smears to identify malarial parasites

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text