Thermo Scientific Chemicals

Trypan Blue, dye content >60%

Catalog number: A18600.14
25 g, Each
Thermo Scientific Chemicals

Trypan Blue, dye content >60%

Catalog number: A18600.14
25 g, Each
Quantity
Catalog number: A18600.14
also known as A18600-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
72-57-1
IUPAC Name
tetrasodium (3Z)-5-amino-3-[2-(4'-{2-[(2Z)-8-amino-1-oxo-3,6-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Molecular Formula
C34H24N6Na4O14S4
InChI Key
WMYVHJWZUUEZNE-ARWFNKCKSA-J
SMILES
[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1\C(=O)C2=C(N)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2\C(=O)C3=C(N)C=C(C=C3C=C2S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1
Appearance (Color)
Dark purple to dark brown or black
Form
Powder
Assay (unspecified)
≥60% dye content
Loss on Drying
≤15.0% (200?C, constant weight)
lambda-max (UV-Vis)
602-610 nm (methanol)

Description

Trypan blue is commonly used in microscopy (for cell counting) and in laboratory mice for assessment of tissue viability. Used in dye exclusion studies in cell culture. A viability dye for quantifying live versus dead cells with permeable membranes, blue. It is also used to identify islet cell cluster cell viability from caprine pancreatic samples. It is also useful to observe hyphea of fungi and Stramenopiles. Trypan blue solution may be used in trypan blue based cytotoxitiy and proliferation assays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trypan blue is commonly used in microscopy (for cell counting) and in laboratory mice for assessment of tissue viability. Used in dye exclusion studies in cell culture. A viability dye for quantifying live versus dead cells with permeable membranes, blue. It is also used to identify islet cell cluster cell viability from caprine pancreatic samples. It is also useful to observe hyphea of fungi and Stramenopiles. Trypan blue solution may be used in trypan blue based cytotoxitiy and proliferation assays.

Solubility
Soluble in water (10 mg/ml at 25°C), ethanol (0.6 mg/ml), and EGME (20 mg/ml).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text