Thermo Scientific Chemicals

4-Iodo-2-nitrotoluene, 97%, Thermo Scientific Chemicals

Catalog number: A18607.06
5 g, Each
Thermo Scientific Chemicals

4-Iodo-2-nitrotoluene, 97%, Thermo Scientific Chemicals

Catalog number: A18607.06
5 g, Each
Quantity
Catalog number: A18607.06
also known as A18607-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
41252-97-5
IUPAC Name
4-iodo-1-methyl-2-nitrobenzene
Molecular Formula
C7H6INO2
InChI Key
QLMRDNPXYNJQMQ-UHFFFAOYSA-N
SMILES
CC1=CC=C(I)C=C1[N+]([O-])=O
Appearance (Color)
Orange to brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
56.5-62.5?C

Description

4-Iodo-2-nitrotoluene is used to produce 3-(4-methyl-3-nitro-phenyl)-1H-indole at the temperature of 40°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodo-2-nitrotoluene is used to produce 3-(4-methyl-3-nitro-phenyl)-1H-indole at the temperature of 40°C.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text