4-Iodo-2-nitrotoluene is used to produce 3-(4-methyl-3-nitro-phenyl)-1H-indole at the temperature of 40°C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Iodo-2-nitrotoluene is used to produce 3-(4-methyl-3-nitro-phenyl)-1H-indole at the temperature of 40°C.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
RUO – Research Use Only
Andrew C. Peacock.; C. Wesley Dingman. Resolution of Multiple Ribonucleic Acid Species by Polyacrylamide Gel Electrophoresis. Biochemistry. 1967, 6 (6), 1818-1827.
C. Wesley. Dingman.; Andrew C. Peacock. Analytical studies on nuclear ribonucleic acid using polyacrylamide gel electrophoresis. Chem. Commun. 1968, 7 (2), 659-668.