Sodium tin(IV) oxide trihydrate is used in the preparation of boscage-like stannic oxide by facile one-step hydrothermal route.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium tin(IV) oxide trihydrate is used in the preparation of boscage-like stannic oxide by facile one-step hydrothermal route.
Solubility
Soluble in water. Insoluble in ethanol and acetone.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
Zhao, K.; Du, G.; Qin, G.; Liu, Y.; Zhao, H. Facile synthesis of boscage-like SnO2 nanorods by hydrothermal method. Mater. Lett. 2015, 141, 351-354.
Zhao, K.; Du, G.; Luo, L.; Qin, G.; Jiang, Q.; Liu, Y.; Zhao, H. Novel multi-layered SnO2 nanoarray: self-sustained hydrothermal synthesis, structure, morphology dependence and growth mechanism. CrystEngComm 2015, 17 (9), 2030-2040.