Thermo Scientific Chemicals

4-Methoxytrityl alcohol, 94%, Thermo Scientific Chemicals

Catalog number: A18620.09
10 g, Each
Thermo Scientific Chemicals

4-Methoxytrityl alcohol, 94%, Thermo Scientific Chemicals

Catalog number: A18620.09
10 g, Each
Quantity
Catalog number: A18620.09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
847-83-6
IUPAC Name
(4-methoxyphenyl)diphenylmethanol
Molecular Formula
C20H18O2
InChI Key
WCRRRAKYYPJJMP-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=C1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
73.0-86.0?C
Appearance (Color)
White
Assay (Silylated GC)
≥92.0%

Description

It is used as an intermediate for preparation of TT-0011.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate for preparation of TT-0011.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text