2-Azacyclononanone has been used as an internal standard liquid chromatography-ultraviolet detection method for the determination of caprolactam in the aqueous food stimulant 3% acetic acid (w/v).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Azacyclononanone has been used as an internal standard liquid chromatography-ultraviolet detection method for the determination of caprolactam in the aqueous food stimulant 3% acetic acid (w/v).
Solubility
Soluble in methanol (almost transparency).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
Bustos J, et al. Migration of e-caprolactam from nylon cooking utensils: validation of a liquid chromatography-ultraviolet detection method. Eur. Food Res. Technol. 2009, 230 (2), 303-313.
Garrido Pedrosa AM, et al. Lanthanide Trifluoroacetate Complexes with 2-azacyclononanone. Thermal and kinetic studies. Therm. Anal. Calorim. 2002, 67 (2), 397-402.