Methyl chlorodifluoroacetate, 98%, Thermo Scientific Chemicals
Methyl chlorodifluoroacetate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl chlorodifluoroacetate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
250 g
Catalog number A18623.09
also known as A18623-09
Price (USD)/ Each
34.50
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
34.50
Add to cart
Methyl chlorodifluoroacetate, 98%, Thermo Scientific Chemicals
Catalog numberA18623.09
Price (USD)/ Each
34.50
-
Add to cart
Chemical Identifiers
CAS1514-87-0
IUPAC Namemethyl 2-chloro-2,2-difluoroacetate
Molecular FormulaC3H3ClF2O2
InChI KeyAWUPLMYXZJKHEG-UHFFFAOYSA-N
SMILESCOC(=O)C(F)(F)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3485-1.3525 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only