Thermo Scientific Chemicals

DL-2-Hydroxybutyric acid sodium salt, 97+%, Thermo Scientific Chemicals

Catalog number: A18636.03
1 g, Each
Thermo Scientific Chemicals

DL-2-Hydroxybutyric acid sodium salt, 97+%, Thermo Scientific Chemicals

Catalog number: A18636.03
1 g, Each
Quantity
Catalog number: A18636.03
also known as A18636-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5094-24-6
IUPAC Name
sodium 2-hydroxybutanoate
Molecular Formula
C4H7NaO3
InChI Key
MOSCXNXKSOHVSQ-UHFFFAOYNA-M
SMILES
[Na+].CCC(O)C([O-])=O
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥97.0 to ≤103.0%
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
≤3.0%
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text